missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human PRKCI (PKC iota), His Tag Recombinant Protein

Human PRKCI (PKC iota), His Tag Recombinant Protein

Brand:  Thermo Scientific™ PV3607

 View more versions of this product

Product Code. 10605203

  • £6900.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • The PKC family of serine/threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways
  • In humans, at least 12 different PKC polypeptides have been identified
  • These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity
  • PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NP_002731
71.5 kDa
PRKCI (PKC iota)
Human
His Tag
Ser/Thr Kinases: AGC Group
Baculovirus
Frozen
Kinase Assay
1 mg
PKC Kinase Family
Recombinant
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Full Length
Drug Development
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers