missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human PRKCI (PKC iota), His Tag Recombinant Protein
Human PRKCI (PKC iota), His Tag Recombinant Protein
Brand: Thermo Scientific™ PV3186
This item is not returnable.
View return policy
Description
- The PKC family of serine/threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways
- In humans, at least 12 different PKC polypeptides have been identified
- These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity
- PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent
Specifications
NP_002731 | |
5584 | |
20 μg | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser/Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Zin-LYTE™, Omnia™ Bulk Substrate | |
Protein kinase C, iota | |
Kinases & Inhibitors | |
Serine/Threonine Kinases |
Kinase Assay | |
71.5 kDa | |
PRKCI (PKC iota) | |
Human | |
His Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
PRKCI | |
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction |