missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PRKG1 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16871255
|
Proteintech
21646-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16861255
|
Proteintech
21646-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphorylates PPP1R12A.Specifications
PRKG1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P0C605, Q13976 | |
19091, 54286, 5592 | |
PRKG1 Fusion Protein Ag16333 | |
Primary | |
-20°C | |
PRKG1 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
cGK 1, cGK1, CGKI, cGKI alpha, cGKI BETA, PGK, PKG, PRKG1, PRKG1B, PRKGR1A, PRKGR1B, Protein Kinase G1 alpha | |
Prkg1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title