Learn More
Gilson™ Pro Pipette Service for Single Channel Pipettes
Product Code. 17298379 Shop All Gilson Products
Click to view available options
Product Type:
Preventive Maintenance and Calibration
Service Offering:
As-found/After Repair Calibration
Calibration Points:
5 x 3 Measurements
Unit Size:
Each
Product Code. 17298379
Brand: Gilson™ SVC101151
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Gilson Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • 5 day turn around time
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Single-channel Pipette
Certifications/Compliance ISO 17025 Accredited Service
Description Data collected before and after repair
Calibration Points 5 x 3 Measurements
Duration 5 Days
Calibration Type Gravimetric
Product Type Preventive Maintenance and Calibration
Product Line Pipette Pro
Service Offering As-found/After Repair Calibration