Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Staining Kits
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards.
£194.00 - £560.00
Specifications
Detection Location | In-Gel Detection |
---|---|
Detection Method | Fluorescence |
Label or Dye | Pro-Q Diamond |
Product Type | Phosphoprotein Gel Staining Kit |
Product Line | PRO-Q™ |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10184652
|
Invitrogen™
MPP33301 |
Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 200 mL |
£194.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
10195032
|
Invitrogen™
MPP33300 |
Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 1 L |
£560.00
1 set |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. The PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.Features include:
• Sensitive—detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis
• Built-in controls—molecular weight marker allows for approximation of protein size and serves as positive and negative control with unique mixture of phosphorylated and non-phosphorylated proteins
• Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitation
Specifications
In-Gel Detection | |
Pro-Q Diamond | |
PRO-Q™ | |
Proteins (Phosphoproteins) |
Fluorescence | |
Phosphoprotein Gel Staining Kit | |
1 Kit |