missing translation for 'onlineSavingsMsg'
Learn More

Probenecid, 98%

Product Code. 11420461
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
10 g
missing translation for 'unitSize'
10g
2.5g
This item is not returnable. View return policy

Product Code. 11420461

missing translation for 'mfr': Thermo Scientific Alfa Aesar B20010.88

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 57-66-9 | C13H19NO4S | 285.36 g/mol

It is a uricosuric drug, that is, it increases uric acid excretion in the urine. It is primarily used in treating gout and hyperuricemia. Probenecid was developed as an alternative to caronamide to competitively inhibit renal excretion of some drugs, thereby increasing their plasma concentration and prolonging their effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (partly miscible), solutions of alkali hydroxides,, ethanol (100 mM), acetone, chloroform, sodium hydroxide (1 M), DMSO (100 mM), methanol, dilute acids (practically insoluble), dilute alkalies, and ether (slightly soluble).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.36
MDL Number MFCD00038402
InChI Key DBABZHXKTCFAPX-UHFFFAOYSA-N
Synonym probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
PubChem CID 4911
ChEBI CHEBI:8426
IUPAC Name 4-(dipropylsulfamoyl)benzoic acid
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 198°C to 199°C
Assay Percent Range 0.98
Quantity 2.5 g
Merck Index 14,7754
Solubility Information Soluble in water (partly miscible),solutions of alkali hydroxides,,ethanol (100 mM),acetone,chloroform,sodium hydroxide (1 M),DMSO (100 mM),methanol,dilute acids (practically insoluble),dilute alkalies,and ether (slightly soluble).
Formula Weight 285.36
Percent Purity 98%
Chemical Name or Material Probenecid
compliance-icons
Product Identifier
  • Probenecid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Probenecid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.