Learn More
Cytiva ProbeQuant™ G-50 Micro Columns
For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50μL. Cytiva Life Sciences™ ProbeQuant™ G-50 Micro Columns use spin-column chromatography.
Brand: Cytiva 28-9034-08
178.43 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Description
- Prepacked with Sephadex™ G-50 DNA grade, and designed for use in a microcentrifuge
- Requires less than four minutes from sample application to collection of purified product
- A portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation
- Tested in nickase, single and double-stranded exonuclease, and RNase assays
Order Info
Conveniently prepacked with Sephadex G-50 DNA Grade and pre-equilibrated in STE containing 0.15% Kathon™ CG/ICP Biocide
Specifications
Rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50μL using spin column chromatography | |
Column | |
50 Pack |
Safety and Handling
- ||Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
- MIXTURE LIST-Contains : reaction mass of: 5-chloro2-methyl-4-isothiazolin-3- one ¢ECno. 247-500-7! and2-methyl-2H-isothiaz
Your input is important to us. Please complete this form to provide feedback related to the content on this product.