Learn More
Invitrogen™ ProcartaPlex™ Cell Lysate Assay Diluent (1X)
ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel for dilution of cell lysate samples.
Brand: Invitrogen™ EPX-11114-000
Additional Details : Weight : 0.01000kg
Description
ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel (Cat. No. EPX060-15823-901) for dilution of cell lysate samples. Diluent is included with the panel, but is also offered here as a stand-alone item.
Reported application
- Multiplex immunoassay
Specifications
Diluent | |
ProcartaPlex™ | |
Wet Ice |
10 mL | |
Store in refrigerator (2°C to 8°C). |
For Research Use Only. Not for use in diagnostic procedures.