Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human Factor X Simplex
The Human Factor X Simplex ProcartaPlex Kit measures Factor X protein and includes all reagents needed to run the assay.
Brand: Invitrogen™ EPX01A-12152-901
Code : NEW
Additional Details : Weight : 0.23750kg
Description
- The Human Factor X Simplex ProcartaPlex Kit measures Factor X protein and includes all reagents needed to run the assay, eliminating the need to purchase a basic kit
- This kit cannot be combined with other simplex kits or panels
- ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument
- ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant,
- Analyzed results can be obtained in as little as four hours
- Store kit at 2-8°C
Kit Contents
- 1 vial Standard Mix
- 1 vial Bead Mix (1X)
- 1 vial Biotinylated Detection Antibody Mix (1X)
- 2 vials Coagulation Control Plasma
- 1 bottle Reading Buffer (1X)
- 1 bottle Wash Buffer (10X)
- 1 bottle Streptavidin-PE (1X)
- 1 bottle Universal Assay Buffer (1X)
- 1 bottle Detection Antibody Diluent (1X)
- 8-tube strip
- Adhesive film
- Flat bottom 96-well plate, black
Specifications
96 Tests | |
Multiplex Immunoassay | |
Plasma (Citrat, EDTA): 25 μL |
Human | |
Plasma (EDTA, Citrate) | |
F10, FX, FXA |