Learn More
Invitrogen™ ProcartaPlex Human High Sensitivity Basic Kit
Assess multiple protein biomarkers in a single sample. Our ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, and growth factors. This kit includes all non-target-specific reagents.
Brand: Invitrogen™ EPXS010-10420-901
Code : NEW
Additional Details : Weight : 0.23750kg
Includes:
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent Human
- Amplification Reagent I
- Amplification Reagent II
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- Explore our broad portfolio of pre-configures panels and combinable single analytes
- For combination of only Simplex Kits an additional Basic Kit is required
- Not intended for human or animal therapeutic or diagnostic use
- Assay diluent, wash buffers, SA-PE, amplification reagents and plates
- This kit does not contain a Multiscreen Filter Plate
Specifications
High Sensitivity Simplex Kit | |
Human | |
See Certificate of Analysis | |
Luminex™ Instruments |
96 Tests | |
Combinable (High Sensitivity Only) | |
High Sensitivity Simplex Kit | |
ProcartaPlex™ |
For Research Use Only. Not for use in diagnostic procedures.