Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human PD-1 (CD279) Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX01A-12214-901
Product Code. 15552277
  • £221.00 / 96 tests
Explore more special offers
Includes:
  • concentrated Magnetic Beads coated with monoclonal antibody to human PD-1 (CD279)
  • human PD-1 (CD279) Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human PD-1 (CD279) monoclonal antibody
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Cell Culture Supernatant, Plasma, Serum
Programmed cell death protein 1, PDCD1
Human
CD279 (PD-1)
Serum, Plasma: 25 μL; CCS: 50 μL