Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human Th9/Th17/Th22/Treg Cytokine Panel (7 plex)
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX070-10817-901
Code : NEW
Additional Details : Weight : 0.23750kg
Includes:
- Human Standard Mix A
- Human Bead Mix C
- Human Detection Antibody Mix C
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit does not contain a Multiscreen Filter Plate
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum |
Human | |
IL-10, IL-17A, IL-21, IL-22, IL-23, IL-27, IL-9 | |
Serum, Plasma: 25 μL; CCS: 50 μL |