Learn More
Invitrogen™ ProcartaPlex™ Microplate Lid
Description
This reusable microplate lid is ideal for use with all ProcartaPlex Luminex protein assays.
This lid is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the 96-well flat bottom plate (Cat. No. EPX-88182-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
- ProcartaPlex Flat Bottom Plate, 96-well (Cat. No. EPX-88182-000)
Specifications
Specifications
| Color | Black |
| Material | Polystyrene |
| Sterility | Non-sterile |
| No. of Wells | 96 |
| Quantity | 96 tests |
| Product Type | Plate |
| Fit for Purpose | Certified Quality |