Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse ENA-78 (CXCL5) Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-26043-901
Code : NEW
Additional Details : Weight : 0.23750kg
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to mouse ENA-78
- mouse ENA-78 Standard (lyophilized)
- mouse Biotin-Conjugate (50x) anti-mouse ENA-78 monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
SCYB5 |
Mouse | |
Cell Culture Supernatants, Serum, Plasma |