Learn More
Invitrogen™ eBioscience™ ProcartaPlex NHP TGF alpha Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-42139-901
Code : NEW
Additional Details : Weight : 0.23750kg
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to non-human primate TGF alpha
- non-human primate TGF alpha Standard (lyophilized)
- Biotin-Conjugate (50x) anti-non-human primate TGF alpha monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
Transforming growth factor alpha |
Monkey | |
TGF-alpha | |
Serum, Plasma: 25 μL; CCS: 50 μL |