missing translation for 'onlineSavingsMsg'
Learn More

Contec™ ProChlor Sporicidal SATWIPES™
BIOPHARMA

Point-of-use SATWIPE system, each double bag pack contains a canister of 120 x 34 gsm melt blown polypropylene wipes and 700 mL ProChlor in a 1 L bottle

Brand:  Contec™ FSWPP02PC

399.99 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 16439565

  • £663.00 / Pack of 1200
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Impregnated wipes at point-of-use meets requirements of Aseptic Transfer Guidelines
  • Blend of hypochlorous acid in purified water achieves a 100% kill against spores in under 1 min.
  • Efficacious against bacteria, fungi, molds, yeasts and spores
  • 0.2 micron filtered and filled in cleanroom environment, ideal for life science cleanrooms
  • Low residue contains no quaternary ammonium compounds or surfactants
  • Fast acting, saves time spent on biodecontamination, can be used for transfer disinfection
  • Double bagged packed in linear tear packaging for easy transfer into cleanroom environment
  • Not classed as corrosive, can be used safely in all areas of cleanroom
  • No hazard classification means only basic PPE required, no special disposal required
  • 120 polypropylene wipes in a canister reduces amount of packaging generated
  • Designed for disinfection of hard surfaces in cleanrooms and controlled environments
Specifications

Specifications

Sporicidal Cleanroom Wipe
200 mm
150 mm
34 gsm
Do not freeze, store below 40°C
Wipes to saturate at point- of-use
10 Canisters/Cs.
Non-sterile
7.9 in.
5.9 in.
Polyester Cellulose
Canister
15 x 20 cm (6 x 8 in.)
Hard surface disinfectant in life science cleanrooms
2000 ppm stabilized hypochlorous acid
Canister of 120 x 34 gsm melt blown polypropylene wipes, 700 mL ProChlor in a 1 L bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers