Learn More
Bel-Art™ ProCulture™ Cordless Homogenizer Unit
Cordless homogenizer unit is hand-held and is ideal for thorough and faster processing of samples.
Brand: Bel-Art™ F65100-0000
99.48 GBP valid until 2025-12-31
Use promo code "25045" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hand-held, cordless homogenizer features a simple push-in, pull-out pestle exchange and is ideal for thorough and faster processing of one-to-many samples
- Optional ProCulture™ Matched Pestles and Tubes assure tight tolerances and thorough homogenization and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension; F65000-0005, F65000-0006, F65000-9006
- Optional ProCulture™ Pestles have a unique ribbed shaft that provides precise finger control of homogenization for shear sensitive samples; also fit most motorized mixer adapters; F65000-0001, F65000-0002, F65000-9001, F65000-9002
Specifications
Cordless Homogenizer Unit | |
CE | |
Homogenizer with Cordless Motor, AA Batteries (2), and Pestle Adapter |
0/1 V | |
Yes | |
Yes |
Product Content Correction
Product Title
WARNING: This product can expose you to chemicals which are known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Spot an opportunity for improvement?Share a Content Correction