Description
- Uniform thermal characteristics
- High heat conductivity
- No wetted part
- Sample equilibration in same tray
- No. of gels: 1 to 12
Specifications
Ettan IPGphor 3 Isoelectric Focusing Unit | |
Ceramic | |
Complete packsize |
IPGphor Manifold Ceramic Tray | |
1 to 12 |