man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Specialty Lab Glassware
Reaction Kettles
10555091
Print
IKA™ LR 2000.4 Reactor Vessel
Reactor Vessel; For Use With LR 2000 P
Brand:
IKA™ 0003064900
Additional Details :
Weight :
0.50000kg
Product Code.
10555091
£9540.00
/ Each
Quantity
Estimated Shipment: 01-10-2024
Log in
to see stock.
Add to basket
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Description
Description
Specifications
Specifications
For Use With (Equipment)
For LR 2000 P
Product Type
Reactor Vessel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers