Learn More
Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts
Use these tilting half-moon blades for stirring in round bottom flasks with ground joint necks. Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts offer tangential flow with little turbulence and universal chemical resistance.
Brand:  Bohlender™ C376-08
 View more versions of this product
Product Code. 10635921
  • £380.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PTFE-jacketed stainless steel shaft with tilting half-moon stirrer blade
  • Double-sided groove and access for the stirrer shaft completely made of PTFE
  • Temperature resistance: -200 to +250°C
  • Blades are available separately and can be mounted additionally
Specifications
Specifications
8 mm
450 mm
29/32
The tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint necks.
1 Each
Moon Shaped Stirrer Shaft