Learn More
Cytiva INV-907 Injection Kit
Range of manual and automated valves to meet variety of performance requirements.
Brand:  Cytiva 18-1110-89
Product Code. 11390152
  • £414.00 / Each
Edge
Explore more special offers
Description
Description
  • Designed for biocompatibility
  • Each valve may be inserted into flow path without causing unnecessary mixing or sample dilution
  • Includes fill port and needle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Injection Kit
ETFE, Titanium
Kit includes fill port and needle.