Description
- Complete 2D analysis - a one-tank solution for 2D electrophoresis using tube gels for the 1st dimension
- Comprises cooled vertical tank complete with casting base and accessories plus the 1D module and accessories
Specifications
Supercooled tank, lid, 2 x notched glass plates, 2 x plain glass plates with 1.5mmthick bonded spacers, dummy plate, 2 x 1.5mm (24 sample) combs, gel extractionplatform, internal gel capillary module, 10 x capillary tubes, 10 x blanking ports and2 x 2D comb (Maxi gel units 205mm x 200mm) |