missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Semi-Dry Blotters
Ideal for fast transfer of proteins and nucleic acids without the need for gel cassettes and tanks.
Brand: Fisherbrand™
Description
- Designed to produce an extremely uniform system for the transfer of proteins and nucleic acids.
- Very low buffer volume and current, 0.8mA per cm2 of gel, produces very little heat, preventing band distortion
- Transfers can be achieved in less than one hour, depending on the molecular weight of the molecules
- Either single gels or multiple gel stacks can be used with the units
- Colour coded electrodes: One electrode is manufactured from stainless steel and the other from platinised titanium, making them virtually corrosion free
- Fast transfer times
- Available in two sizes
V20-SDB: 20 x 20cm format - accommodates up to four 10 x 10cm mini-gels or one 20 x 20cm maxi-gel
Specifications
Semidry Blotter | |
7.5 x 27 x 27 cm | |
80 mA / 0.8 mAcm-2 |
Active Transfer Area: L x W: 10 x 10 cm | |
Recommended Power Supplies: EV265, 261 and 215 |