missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Inverso TC100 Inverted Biological Microscope with Fluorescence

An ideal microscope for the typical working laboratory, built to a high quality specification.

Brand:  Medline Scientific™ 3660.8000M


Code : 50

Additional Details : Weight : 0.10000kg

Product Code. 12653025

  • £13395.00 / Each
Estimated Shipment: 06-11-2024
to see stock.
Add to basket

Description

Description

  • Robust enough to withstand the rigors of constant daily use
  • Ideal for multiple usage as there is no manual adjustment
  • Trinocular optical head, inclinded at 45°
  • HB0 100W Ultra Hi-voltage Spherical Mercury Lamp
Specifications

Specifications

1 spare bulb, 1 spare fuse, allen key for condenser locking, dust cover and instruction manual
N.A.0.3 (removable)
Brightfield Plan Achromatic
220/240 V
WF 10X/22 mm
Trinocular, 45° Inclined
220/240 V
Field diaphragm
Quadruple revolving with click stop, Inverted
50/60 Hz
HB0 100w Ultra Hi-voltage Spherical Mercury Lamp
Inverso
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers