missing translation for 'onlineSavingsMsg'
Learn More

Interscience easySpiral Dilute Automatic Plater and Dilutor

easySpiral Dilute, Automatic Diluter And Plater Serial Dilutions & Standardised Plating 55 mm

Brand:  Interscience 414000


Code : 50

Additional Details : Weight : 16.40000kg

Product Code. 13415199

  • £39965.00 / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

Automatic plater for Petri dishes, Plating modes: exponential (Spiral), Constant (= manual type) circle, pouring. Petri dish diameter: 55 mm, 90 mm or 150 mm. Delivered with:
  • 1 plating ring for dia. 90 mm Petri dishes (by default on the unit) - 1 box with:
  • 1 blue dye for testing (Ref 415 530)
  • 1 syringe with silicone tubing
  • 1 plating ring for dia. 55 mm Petri dishes
  • 1 plating ring for dia. 150 mm Petri dishes
  • 1 Spiral™ counting grid for dia. 90 mm Petri dishes (Ref 413 014)
  • 1 Circle mode counting grid for dia. 90 mm Petri dish (Ref 413 026) 1 set of 2 filtres (Ref 413 009)
  • 1 software (minimum requirements: screen resolution of 1280 x 960) 1 USB cable A/B
  • 1 Circle mode counting grid for dia. 150 mm Petri dish (Ref 413 027)
  • 1000 Spiral™ DB50 beakers (Ref 415 100)
  • 1 EnzyClear™ liquid detergent (Ref 413 029)
  • 4 single sets of GL 45 bottles connectors (Ref 413 003)
  • 1 double set of GL 45 bottles connectors (Ref 414 004)
  • 1 stylus (Ref 414 001)
  • 1 power cable
  • 1 user manual
  • 1 EnzyClear™ quick user guide
Specifications

Specifications

Automatic Dilutor and Plater
Not Autoclavable
dilutions from 10^-1 to 10^-5
easySpiral Dilute platers are used for all types of microbiological analyses in the food, pharmaceutical, cosmetic and public research institutes ...
easySpiral Dilute
Plating in 134 seconds (disinfection + 5 dilutions + 1 plating of a dilution). PATENTED
16.57 in.
8.60 lb
100-240 V - 50-60 Hz
45 W
50-60 Hz
± 0.5% (in normal mode)
ISO 7218, ISO 4833-2, FDA BAM (Bacteriological Analytical Manual) and AOAC 977.27
50, 100, 200 μL
Delivered with: 1 turntable ring for dia. 90 mm Petri dishes, 1 blue dye for testing, 1 syringe with silicone tubing, 1 plating ring for dia. 55 mm Petri dishes, 1 plating ring for dia. 150 mm Petri dishes, 1 Spiral counting grid for dia. 90 mm Petri dishes, 1 circle mode counting grid for dia. 90 mm Petri dish, 1 set of 2 filters, 1 software, 1 USB cable A/B, 1 Spiral counting grid for dia. 150 mm Petri dishes, 1 circle mode counting grid for dia. 150 mm Petri dish, 1000 DB50 beakers, 1 EnzyClear liquid detergent, 4 connection kits for GL45 bottle, 1 double kit for GL45 bottles, 1 stylus, 1 power cable, 1 user’s manual, 1 EnzyClear quick user guide.
2%
39.4 x 42.1 x 28.9 cm
11.38 in.
16.6 kg
100/240 V
T1.6A L 250V 5 x 20 mm
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers