missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 7750 KF Module Titrator with 10 mL Exchangeable Head

Automatic Titrator for exchangeable heads for both potentiometric titration and volumetric KF titration. The TitroLine™ 7750 combines the features of the potentiometric titrator TitroLine™ 7000 and the volumetric Karl Fischer titrator TitroLine™ 7500.

Brand:  SI Analytics™ 285220940


Additional Details : Weight : 6.00000kg

Product Code. 15514923

  • £7695.00 / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket

Description

Description

  • High visibility, full color display easily viewed from distance and extreme angles
  • Reagent data securely stored in intelligent and interchangeable modules
  • Includes up to three USB, one LAN and two RS232 ports for expansion and connection of devices
  • Export results as PDF or CSV, also to networks
  • Automatic wireless recognition of SI Analytics ID electrodes guarantee accurate calibration and measurements
  • High resolution pH/mV-electrode and temperature inputs for pH, ISE, redox (ORP) or photometric titrations
  • Perfect for non-aqueous titration such as acid and base numbers in oils (TAN and TBN)
  • Polarisable electrode input for set endpoint titrations (Dead-stop + KF)
  • Linear (fixed increment) and dynamic equivalence point titration mode and titration to endpoints
  • pH Stat titrations
  • Volumetric KF titrations
  • Single and continuous measurement
  • Formula editor for individual calculations
  • User management/password
  • Titrations with new photometric sensor OptiLine 6
  • Dosing with up to two connected piston burettes per method
  • Titration with external piston burette
Specifications

Specifications

EMC compatibility according to the Council Directive: 2004/108/EG; applied harmonized standards: EN 61326-1:2006; Low-voltage directive according to the Council Directive 2006/95/EG Testing basis EN 61 010, Part 1
Basic unit with 10 exchangeable head, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1770, micro double platinum electrode KF 1100, starter kit, power supply 100 to 240 V
Polypropylene Housing, Polyester Coated Keyboard
-2000 to +2000 to 0.1 ± 1
Max. 80% for temperatures up to 31°C, linear decrease down to 50% relative humidity at a temperature of 40°C
50 to 1000 U/min.
10°C to 40°C
100 to 240 V, power input: 30 VA
3 x USB (2 x USB-A and 1 x USB-B), 2 x RS-232-C and 1 x LAN
pH/mV-input with 24 bit transducer. Electrode socket according to DIN 19 262 or BNC with RFID receiver for SI Analytics ID electrodes
100 to 240 V; 50/60 Hz, power input: 30 VA
-3.0 to 18.00 to 0.002 ± 1
20.000 steps
Pt 1000 to 75°C to 95°C / NTC 30 KOhm -40°C to +125°C
250 x 80 x 130 mm (height without stand rod)
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers