missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ K-PHOENIX-90 Combination Safety Storage Cabinets with Door Open Arrest System

For the safe and separated storage of aggressive acids, alkalis and flammable hazardous substances in working areas.

Brand:  Asecos™ 3033300133839


3535.51 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 573.00000kg

Product Code. 15526865

  • £4768.94 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Hermetically sealed storage compartments with fire-resistant dividing walls
  • Integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with EN 14470-1 and TRGS 510
  • Robust construction and longevity
  • Triple-hinged doors, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for protection against corrosion
  • Door open arrest system as standard for the convenient and safe removal or storage of containers
  • Doors lockable with profile cylinder with integration into an existing locking system possible
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • Fire resistance of 90 minutes
  • Easy alignment – adjusting aids to compensate for uneven floors
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

6 x 25 kg/5 L sheet steel drawers; 6 x 25 kg/11 L PP pull-out shelves with PP-sump
Storage of aggressive and flammable hazardous materials
Steel
617 x 1197 x 1969 mm
Light Gray
Wing
EN 14470-1, TRGS 510
Safety storage cabinet
Powder-coated
Door open arrest system
575 kg
Drawers: 25 kg/5 L each, Shelves: 25 kg/11 L each, Max load: 600 kg
3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers