missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ C40 Cuvette UV-VIS Spectrophotometer

For cuvette applications. Suitable for all common photometric measurements like protein assays, kinetics and standard curve. Intuitive touchscreen operation, small footprint and network integration for convenient lab bench operation. Recalibration-free patented technology.

Brand:  Implen™ C40-Mobile


Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 15528783

  • £7056.00 / Each
Estimated Shipment: 30-08-2024
to see stock.

Description

Description

The Implen NanoPhotometer™ C40 UV/VIS spectrophotometer offers full scan data from 200 - 900 nm for all cuvette based photometric applications in Life Sciences. It can be upgraded to NanoVolume using the Implen Submicoliter Cell. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.

No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.

  • Detection range dsDNA 0.1 ng/μL - 130 ng/μL
  • Detection range BSA 0.003 mg/mL - 3.7 mg/mL
  • Photometric Range 0 - 2.6 A
  • Scan range 200 - 900 nm
  • Center Height (Z-Height) 8.5 mm
  • Suitable for all cuvettes with outside dimensions 12.5 x 12.5 mm
  • Temperature controlled cuvette port 37°C
  • Measure time for full scan range: 2.5 seconds
  • Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
  • Recalibration-free due to True Path Technology™
  • Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
  • Control options: Onboard with built-in 7” Touchscreen, Computer and Tablet
  • In and Output Ports 2x USB A, USB B, HDMI, Ethernet, Wi-Fi
  • 21 CFR part 11 compliant software (optional)
  • Built-in file server
  • LIMS integration through REST API
  • Built-in battery pack, operation time: up to 8 h
  • Made in Germany
Specifications

Specifications

< 1.75 % at 0.7 A (0.67 mm path) at 280 nm of the reading
8.5 mm
3648-element CCD Array
200 x 200 x 120 mm
Xenon Flash
Spectrophotometer
200 to 900 nm
Submicroliter Cell
2.5 sec
37 °C ± 0.5 °C
Linux based NPOS
± 0.75 nm
0 to 2.6 A
2x USB A, USB B, HDMI, Ethernet, WiFi
7 in. Glove Compatible Touchscreen, 1024 x 600 pixels
English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
<1.8 nm
90/250 V
Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom
Battery, Power Supply, USB Cable, Dust Cover, Software and Manual
1
10, 5, 2, 1, 0.5 mm
Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS
4.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services