Learn More
Spex SamplePrep™ Freezer/Mill™ Dual Chamber Cryogenic Grinder
High-throughput, dual chamber cryogenic grinder with self-contained liquid nitrogen tub and insulated case.
Brand:  Spex SamplePrep™ 6875D-230
Code : NEW
Additional Details : Weight : 42.00000kg
Product Code. 15595546
  • £23455.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Designed for cryogenic grinding of tough and temperature sensitive samples
  • Mill cools samples to cryogenic temperatures
  • Liquid nitrogen required for operation
Specifications
Specifications
Freezer/Mill Dual Chamber Cryogenic Grinder
50.8 x 53.34 x 64.77 cm