missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Freezers

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers offer a sustainable choice for clinical and laboratory storage

Brand:  Thermo Scientific™ TSX2320FV


5229.40 GBP valid until 2024-08-31
Use promo code "21622" to get your promotional price.



Additional Details : Weight : 191.00000kg

 View more versions of this product

Product Code. 15658416

  • £6972.53 / Each
In Stock
to see stock.

Description

Description

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated.   The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. 
 
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 50% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Temperature range of -35° to -15°C, factory setpoint at -20°C
  • Lower HVAC costs as a result of less heat emission into the environment*
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details
Specifications

Specifications

High-Performance Freezer
650 L
Solid
4
Microprocessor
Warranty varies by country, please contact us for details
28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
37.5 in.
95.3 cm
78.6 in.
199.6 cm
28 in.
71.1 cm
Wireless
Smart-Vue Compatible
CEE 7/7
208-230V, 50Hz
Energy efficient
421 lb.
367 lb.
4
23 cu. ft.
-25°C to -15°C (-20°C)
1
UL, cUL, CE
TSX Series
Manual
37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6 cm)
28.5 in.
72.3 cm
58 in.
147.3 cm
24 in.
61 cm
Optional
16 A
230 V
50 Hz
R290
191 kg
166 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.