missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560543
This item is not returnable.
View return policy
Description
Immunogen sequence: CSCCHHSSKE DFKSQSWRQG PDREPESHSP QQGCYPEGPM GIIQHLEPSA HAAGCGKDS Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 31%.
The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. This gene is activated at alkaline pH. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2008].Specifications
KCNK17 | |
Polyclonal | |
Unconjugated | |
KCNK17 | |
2P domain potassium channel Talk-2; Acid-sensitive potassium channel protein TASK-4; K2p17.1; KCNK17; Potassium channel subfamily K member 17; potassium channel, two pore domain subfamily K, member 17; potassium two pore domain channel subfamily K member 17; TALK2; TALK-2; TASK4; TASK-4; TWIK-related acid-sensitive K(+) channel 4; TWIK-related alkaline pH-activated K(+) channel 2; UNQ5816/PRO19634 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96T54 | |
KCNK17 | |
Recombinant protein corresponding to Human KCNK17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only