missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 2%

System starter for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel EX 2% agarose gels.

Brand:  Invitrogen™ G8342ST


Additional Details : Weight : 1.10000kg

Product Code. 15796558

  • £2690.00 / Each
Estimated Shipment: 30-07-2024
to see stock.

Includes: E-Gel Power Snap Electrophoresis device,E-Gel Power Snap Camera,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.

Description

Description

The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 2%, provides everything you need to quickly start using precast E-Gel EX 2% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.

The kit includes:

  • 10 E-Gel EX agarose gels, 2% (Cat. No. G401002)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
  • Gel opening knife (Cat. No. EI9010)

Services

1 year warranty

Specifications

Specifications

• 1 electrophoresis device
• 1 camera device
• 10 E-Gel gels EX 2%
• 1 vial E-Gel 50 bp DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter
• 1 knife

Store at room temperature.
Electrophoresis System Starter Kit
Room Temperature
240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.