missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits

Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.

Brand:  Thermo Scientific™ SPD1030A-230


Additional Details : Weight : 86.00000kg

 View more versions of this product

Product Code. 15916171

  • £18930.72 / Each
Estimated Shipment: 09-10-2024
to see stock.
Add to basket

Description

Description

Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).

Easy to Use:

  • Plug and play installation:. Includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
  • Audible alarm to alert user when operation is finished

Robust, Low-Maintenance Design:

  • Oil-free vacuum pump is virtually maintenance free
  • PTFE coated chamber protects the chamber from corrosion, while making cleaning easy

Enhanced Evaporation:

  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced length vapor path from concentrator to refrigerated vapor trap accelerates evaporation
  • Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates

Solvent and Solvent Combinations:

  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Other organic solvents

SPD1030A and SPD2030A includes:

  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD1030P1 Kit includes:

  • Fully Integrated SpeedVac system
  • 40 × 1.5mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD2030P1 kit

  • Integrated SpeedVac system
  • 200 × 1.5mL rotor (RH200-12)
  • CryoCool™ 1L heat transfer fluid
  • Glass condensation Flask (GCF400)
Specifications

Specifications

<10 Torr (13 mbar, 1.3k Pa)
UL Listed to US and Canadian requirements. CE marked
86 kg
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
PTFE-coated Aluminum
230 V 50 Hz
66 kg
146 lb.
Diaphragm pump, cold trap and vacuum gauge, RH40-11 rotor (40x1.5-2ml tube)
SPD1030
Ambient to 80°C
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor- order separately)
8 A
25 x 26 x 16 in. (64 x 66 x 41 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services