missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Enzyme Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.
Brand: Thermo Scientific™ TSX3020EV
Description
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Every enzyme freezer comes with a complete set of nine shelves with enzyme storage bins to optimize organization and retrieval of smaller materials.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 50% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Temperature range of -35° to -15°C, factory setpoint at -20°C
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models
Spécification
High-Performance Freezer | |
827 L | |
Solid | |
9 | |
Microprocessor | |
Manual | |
28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm) | |
38.9 in. | |
99.0 cm | |
78.5 in. | |
199.4 cm | |
34.0 in. | |
86.4 cm | |
Wireless | |
Smart-Vue Compatible | |
CEE 7/7 | |
208-230V, 50Hz | |
R290 | |
251 kg | |
220 kg |
29.2 cu. ft. | |
-25°C to -15°C (-20°C) | |
1 | |
UL, cUL, CE | |
TSX Series | |
38.9 x 34.0 x 78.5 in. (99.0 x 86.4 x 199.4 cm) | |
38.9 x 34.0 x 78.5 in. (99.0 x 86.4 x 199.4 cm) | |
28.5 in. | |
72.3 cm | |
58.0 in. | |
147.3 cm | |
30 in. | |
76.2 cm | |
Optional | |
15 A | |
230 V | |
50/60 Hz | |
552 lb. | |
484 lb. | |
9 |
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.