missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ G:BOX Chemi XX9 F0.8 Gel Documentation System

G:BOX Chemi XX9 F0.8 Gel Documentation System is a versatile gel documentation system for blots and gels.

Brand:  Syngene™ GBOX-CHEMI-XX9-F08-E


Additional Details : Weight : 50.00000kg

Product Code. 16337545

  • £31275.00 / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

9MP, 16bit camera system with motor driven fixed lens f0.80 with motor driven sample racking system. With 7 position motor driven filter wheel. Max image size 34.5 x 27.6cm. Includes EPI LED white lights and wiring for edge lighting unit (optional). With GeneSys software and unlimited copies of GeneTools. No transilluminator or white light converter supplied. PC required (not included)

High performance imaging has been proven to achieve comparable sensitivity to film with the added benefit of using advanced software tools to optimise the imaging of faint bands. The G:BOX Chemi XX9 is capable of many imaging applications including DNA/Protein gels, chemiluminescence, stain free gels, coloured multiplex fluorescence blots, 2D gels and In Vivo applications (additional accessories may be needed). There is a Peltier cooled, high resolution 9 MP camera, capable of producing 16 bit images (with 65535 grey scales). The camera chip is a leader in the field with a QE of 73% at 425 nm (peak 77%). The system has an F0.8 lens with motorized iris and focus (with feedback) for short acquisition times. The stretch darkroom has a unique motorized lift to utilise zoom features, with autofocus and ensuring the maximum image resolution of the gel or blot is captured. The system as standard comes with a seven position filter wheel, epi white light and an optional LED gantry which gives capability for fluorescence imaging, for red, green, blue and near-IR imaging (additional accessories will need to be purchased).

GeneSys is an application driven software designed for rapid image capture of biological samples. It has a simple icon based user interface usable on Win 10, 8 and 7 platforms. GeneSys allows the user to take the guess work out of imaging, and produce quick and simple publication quality images. All files produced are a unique SGD format, where only raw data is analysed for quantification of samples. Files can also be saved as TIFF, JPEG, BMP formats. User protocols can be saved to review previous images captured, and easily replicate future imaged gels. All captured images can be simply edited with annotations, edit tools, image sharpen and exported to GeneTools.

GeneTools software provides the user with rapid quantification of samples and goes from blots to analysed data in a few quick steps.
Analysis sample types include molecular weight, colony counting, spot blot and band matching.

All samples can be quantified, including multiplexed protein fluorescence analysis of up to 5 samples at once. Raw data files can be quickly quantified to produce true data for publication data. High quality reports can be exported in Word and Excel files for simple review.

  • Camera system with motor driven sample racking system
  • Fixed lens F0.80
  • With 7 position motor driven filter wheel
  • Max image size 34.5 x 27.6 cm
  • Includes EPI LED white lights and wiring for edge lighting unit (optional)
  • With GeneSys software and unlimited copies of GeneTools
  • No transilluminator or white light converter supplied
  • PC required
Specifications

Specifications

None
230 V
7-position Filter Wheel
9 MP
GBOX-CHEMI-XX9-F08
9 MP, 16 bit
Gel Documentation
Gel Documentation System
50 Hz
21.7 in.
38.9 in.
22.4 in.
F0.8
34.5 x 27.6 cm image
Syngene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services