missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ L/S™ Network-Compatible Pump Systems
Networking drives offer either native Ethernet/IP or Profibus communication with no additional hardware or firmware required.
Brand: Masterflex™ 77975-72
Description
- Flow rate 0.001 to 3400 mL/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
- Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
- Remote control available using analog communication—voltage, current and contact closure
- Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
- Antidrip function ensures dispensing accuracy
- Models with open-head sensor stops the drive when the pump head is opened
Spécification
1 | |
LCD | |
0.62 to 3700 mL/min. | |
L/S drive 07575-30, Cytoflow™ pump head 77253-02, 10 ft. C-Flex™ ULTRA™ tubing, and 6 ft. (1.8 m) line cord with IEC 320/CEE22 socket | |
37.03 x 22.86 x 23.65 cm | |
2.2 A at 115 V, 1.1 A at 230 V |
Connections: Profibus, Bluetooth LE, USB | |
IP66 | |
0.1 to 600 rpm | |
Stainless Steel | |
90 to 260 VAC |