Learn More
Syngene™ Airclean 600 Ductless Workstation, ULPA
Ductless workstation equipped with a ULPA filter and AirSafe controller.
Brand:  Syngene™ SYN-32DWASU-E
Additional Details : Weight : 90.00000kg
Product Code. 16504341
  • £6030.00 / Each
Estimated Shipment: 13-09-2024
to see stock.
Description
Description
  • Airclean 600
  • Ductless
  • With AirSafe controller
  • ULPA filter installed
Specifications
Specifications
ULPA
AirSafe Controller
32 in.
81.28 cm