Learn More
Agilent BioTek Barcode Reader Option for Synergy Neo Microplate Reader
For use with Synergy Neo Microplate Readers.
Brand:  Agilent BioTek 1030008
Additional Details : Weight : 1.40000kg
Product Code. 16542002
  • £8540.00 / Each
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
  • Reads 1-D and 2-D barcodes positioned on long or short sides of microplates
Specifications
Specifications
Barcode Reader Option
Synergy Neo Microplate Reader
Reads 1-D and 2-D barcodes positioned on long or short sides of microplates