Learn More
Thermo Scientific™ LabTower™ AFT Clinical Water Purification System
Ideal to feed clinical analyzer systems, these 20 or 40L/hr systems ship complete with pre-treatment and 100L integrated tank.
Brand: Thermo Scientific™ 50135466
Descripción
Ideal to feed clinical analyzer systems, choose either the Thermo Scientific™ LabTower AFT 20 or 40L/hr version and it will ship complete with pre-treatment and 100L integrated tank.
The system utilizes the following water purification technologies:
- Pre-treatment by means of a 10inch mark (") filter housing with a 5Glc mum filter and hardness stabilizer for protection of downstream purification media
- Reverse osmosis module, for the removal of organic and inorganic contaminants, micro organisms, particles and colloids
- Ion Exchanger, for the removal of dissolved organic substances and inorganic ions
- Digital control, for automatic monitoring and controlling of the reverse osmosis system
- Storage tank with water level control for fully automatic maintenance of a stock of high purity water that is protected from light
Especificaciones
0.1 to 1.0 μS/cm | |
50 Hz | |
20 L/hr. | |
LabTower AFT 20 | |
1500 mm | |
2 to 6 bar | |
20 L/hr. | |
2°C to 35°C | |
450 mm |
230 V 50 Hz | |
0.99 | |
580 mm | |
R 3/4 in. | |
66 kg | |
0.25 kW | |
10 to 1 MΩ·cm | |
230 V |