missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Refrigerators - Crated

The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

Brand:  Thermo Scientific™ TSG400RESCW


3243.63 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.



Additional Details : Weight : 113.00000kg

 View more versions of this product

Product Code. 17117758

  • £3794.75 / Each
In Stock
to see stock.

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from 400L to 1500L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
Specifications

Specifications

TSG Series General Purpose Laboratory Refrigerator
2°C to 10°C
R290
1
30 kg
Painted Steel
CE
Digital Microprocessor
12 Months
Crated
Key Lock
493 x 500 x 1405 mm
UK
50 Hz
150 kg
400 L
+5°C
Solid
5 Adjustable
Steel
Painted Steel
Yes
1 Rear Access Port
Adaptive Defrost
Alarm
Smart-Vue Pro Compatible
655 x 600 x 1955 mm
230 V
Energy Efficient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services