missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Flow Cytometer, blue/red/violet6/yellow

Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer

Brand:  Invitrogen™ A29004CFR


Additional Details : Weight : 26.00000kg

Product Code. 17123069

  • POA / Each
Estimated Shipment: 13-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • The 2019 Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT flow cytometer offer adaptable optical configuration options so you can get the most out of your multicolor analysis.
  • Free Attune NxT software with researcher-inspired upgrades
  • New green laser option for expanded utility and fluorochrome options
  • Simple push-button sample recovery function
  • The innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition speeds up to 10X faster than other flow cytometers. This technology reduces the time needed to obtain results.
  • True 35,000 event/sec (+10% coincidence rates)
  • Maximum electronic speed of 65,000 events/sec
  • Analyze small or limited samples faster than ever
  • Efficiently run traditionally long assays
  • The Attune NxT flow cytometer is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, microbiological assays, and more.
  • The instrument’s modular design provides the flexibility to meet future capability needs with a variety of upgrade options. Purchase what you need now and easily upgrade later. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.
  • Flexibility—1 to 4 lasers, 14 colors, and 16 parameters
  • Field upgradeable—choose to add additional lasers when you decide
  • Customizable band pass filters for expanded fluorochrome choices
  • The Attune NxT Flow Cytometer easily runs most samples including cardiomyocytes. The system is designed with a clog-resistant syringe-driven system to push the sample with a force of up to 75 psi.
  • Flexibility in sample concentrations and sample types
  • Elimination of time-consuming concentration steps with No-Wash, No-Lyse Filter Kit
  • Analysis of dilute samples, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
  • Acquisition of a high volume of samples in minimal time


Specifications

Specifications

• Attune Acoustic Focusing Cytometer
• Minitower Windows™ 7 64-bit computer with 23-inch flat panel monitor
• Power cord with adapters
• USB cable
• User manual document CD
• Attune NxT software
• Quick reference card
• 1 vial of Attune Performance Tracking Beads
• 1 bottle of Attune Wash Solution
• 1 bottle of Attune 1X Shutdown Solution
• Attune Focusing Fluid (10 L)

Blue Excitation Laser (488 nm), Red Excitation Laser (638 nm), Violet Excitation Laser (405 nm), Yellow Excitation Laser (561 nm)
Room Temperature
Tube(s), 384-well plate, 96-well plate
Attune™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers