Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System, AB Assurance extended warranty, SmartStart, 384-well
The Applied Biosystems QuantStudio 7™ Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Package includes the QuantStudio 7™ Pro Real-Time PCR System, SmartSmart™ Orientation, and a one-year extended warranty.
Brand:  Applied Biosystems™ A45583
Additional Details : Weight : 50.00000kg
Product Code. 17152106
  • £74743.00 / Each
Estimated Shipment: 27-08-2024
to see stock.
Description
Description
  • Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint
  • The QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
  • Designed for ultimate ease-of-use, the system offers high quality, high throughput, and excellent reliability
  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
  • Space savings with small footprint and true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
  • Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • An optimal user experience for researchers who want to work smarter in the lab
  • Package includes the QuantStudio 7™ Pro Real-Time PCR System, SmartSmart™ Orientation, and a one-year extended warranty
  • Warranty includes comprehensive service coverage and one planned maintenance vist
Services
1 year extended
Specifications
Specifications
Interchangeable
Real Time Thermal Cyclers
12 in. Color Touchscreen
Room Temperature
QuantStudio™ 7 Pro Real Time PCR System (384-well, 0.02 mL block), 1 year extended Warranty, and SmartStart™ orientation
QuantStudio™
Energy Efficient, Fewer Resources used and Less Waste
QuantStudio 7 Pro