Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package
Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint.
Brand:  Applied Biosystems™ A45585
Additional Details : Weight : 50.00000kg Transport : UN number : 3481 Chem class : 9
Product Code. 17172106
  • £75543.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • Includes facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.
  • Provides reproducibility with minimal well-to-well and instrument-to-instrument variation.
  • Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
  • The Connect platform provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the platform are web browser-based software tools that are flexible, fast, and easy to use. These software tools support a variety of analyses including relative quantitation, standard curve, high resolution melt (HRM), genotyping, and presence/absence. They are compatible with Mac or PC as well as most Applied Biosystems instrument file types
  • The system is delivered with one block type; other block types may be purchased separately.
  • In addition, an Orbitor Microplate Mover is available for this system to support high-throughput workflows.
  • Run hundreds of real-time PCR reactions using TaqMan Array 384-well microfluidic cards. And when integrated with the Orbitor Microplate Mover (available separately), the QuantStudio 7 Pro system maximizes your throughput capabilities.
  • Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
  • Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
Specifications
Specifications
Interchangeable
QuantStudio™, ViiA™ 7 System
Touchscreen
Laptop
384-well Plate
QuantStudio™
Room Temperature