Learn More
Invitrogen™ PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi Gel Tank Welcome Pack
The PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack contains: PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank
Brand: Invitrogen™ PSC350TT
Description
The PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack contains:
- PowerEase Touch 350W Power Supply
- SureLock Tandem Midi Gel Tank
About the PowerEase Touch 350W Power Supply
The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.
About the SureLock Tandem Midi Gel Tank
The Invitrogen SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
Specifications
Room Temperature | |
Midi | |
PowerEase | |
4.3 inch backlit LCD | |
One | |
Yes | |
30.3 inch x 23 inch x 10.2 inch | |
300 VDC |
High throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE and western blotting | |
3 A | |
Up to 999 min./step | |
4 (4 positive voltage and 4 negative voltage) | |
Room Temperature | |
One year | |
2.1 kg | |
350 watts |