missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy H1 Hybrid Multi-Mode Reader, IVD

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. For In Vitro Diagnostic Use.

Brand:  Agilent BioTek SH1FG-SI


Additional Details : Weight : 22.50000kg

 View more versions of this product

Product Code. 17237999

  • POA / Each
Estimated Shipment: 14-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »


Description

Description

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. The patented Hybrid Technology offers flexibility and sensitivity across a broad range of applications. The modular platform allows upgrading to expand functionality as your laboratory workflows change

Synergy H1 offers continuously variable bandwidth monochromators for fluorescence excitation and emission wavelength selection. Fluorescence bandwidth can be set between 9 and 50 nm, in 1 nm increments, allowing users to fully optimize reader settings to drive the best assay performance compared to fixed bandwidth systems.

Specifications

Specifications

230 to 999 nm (1 nm increments)
CE and TUV marked. RoHS compliant.
46.4 cm
18.3 in.
11 sec. (96 wells), 22 sec. (384 wells)
0 to 4 OD
13 in.
33 cm
Proprietary Hybrid Technology
2 Syringe Pumps
130 W max.
2.5 to 4 pM monochromators, 0.25 pM filters fluorescence, 20 amol ATP monochromator, 10 amol ATP filter luminescence, 1.2 mP fluorescence polarization, Europium 40 fM with filters, 1200 fM with monos time resolved fluorescence
<55 lb.
14.8 in.
6- to 384-well Plates, Take3 Micro-volume Plates
Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
End Point, Kinetic, Spectral Scanning, Well Area Scanning
Absorbance: ±2 nm; Dispenser: ±1 μL or 2%
0 to 20%
BioStack and Third Party Automation Compatible
4-Zone Incubation to 45°C
Linear, Orbital, Double Orbital
5 to 1000 μL, 1 μL increments
50/60 Hz
Compatible with gas controller; Full-light luminescence; Filter/dichroic fluorescence; Fluorescence polarization; Time resolved fluorescence; Filtered luminescence
H1FG
Multi-Mode Microplate Reader
±0.1% (CO2 and O2)
Fluorescence Intensity Monochromator: 250 to 700 nm (900 nm option), Fluorescence Polarization: 280 to 700 nm (850 nm option)
25 kg
37.5 cm
Photodiode, PMT
Xenon Flash
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers