missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L

The Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.

Brand:  Thermo Scientific™ CE8140BB


Additional Details : Weight : 327.00000kg Transport : UN number : 2800 Chem class : 8

Product Code. 17314650

  • £46122.13 / Each
Estimated Shipment: 16-01-2025
to see stock.

Description

Description

Designed for both liquid and vapor phase storage, the Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.

Specifications

Specifications

Auto-fill LN2 Cryogenic Storage
797 L
Stainless Steel
Stainless Steel
3 in. casters (standard)
42 in.
1067 mm
Access Port for Independent Probe
100/230 V
50/60 Hz
469 kg
327 kg
28.14 cu. ft.
1
Vacuum
CE
49 x 51 x 67 in. (1245 x 1295 x 1702 mm)
17.5 in.
445 mm
0.73A at 110 V and .35A at 230 V
100-230 V
40,600 2 mL vial capacity (max)
720 lb.
Vials (40,600 x 2 mL)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services