missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 1745 L

The Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.

Brand:  Thermo Scientific™ CE8180BB


Additional Details : Weight : 781.00000kg Transport : UN number : 2800 Chem class : 8

Product Code. 17324650

  • £72022.80 / Each
Estimated Shipment: 15-11-2024
to see stock.

Description

Description

Designed for both liquid and vapor phase storage, the Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.

Versatility

  • CryoExtra accepts both vertical and horizontal racks
  • User comfort features and convenient, built-in workspace
  • Designed for vapor-phase storage
  • Folding step for user comfort

Quality

  • Outstanding temperature uniformity
  • Large LED temperature display
  • Lockable lid for sample protection
  • Advanced temperature monitor and alarms
  • Microprocessor-based LN2 level control

Includes:

Built-in battery back-up system: battery power used to maintain power to controller for temperature and level updates during a power outage.

0.5 in. 45° flare, 6′ (1.8m) stainless steel flexible hose

Requires:

22psi low pressure LN2 supply tank (sold separately)

Compatible with:

Ask your customer service representative about racks and accessories for CryoExtra Storage Systems.

Note:

Product height clearance: 77 in. for CryoExtra 20 and CryoExtra 40; 88 in. for CryoExtra 80; 93 in. for CryoExtra 94

Ordering Information:

  • Optional racks, canisters, frames and risers allow you to create an organized inventory system that provides optimum storage in liquid or vapor phase. Contact your customer service or sales representative for details.
  • Warranty varies by country, please contact us for details
Specifications

Specifications

Auto-fill LN2 Cryogenic Storage
1745 L
Stainless Steel
Stainless Steel
3 in. casters (standard)
60 in.
1524 mm
Access Port for Independent Probe
110/230 V
50/60 Hz
2365 lb.
1721 lb.
Vials (80,600 x 2 mL)
61.62 cu. ft.
1
Vacuum
CE
60 x 60 x 68.19 in. (1524 x 1542 x 1732 mm)
25 in.
635 mm
0.73A @110 V and .35A @230 V
100-230V
80,600 2 mL vial capacity (max)
1075 kg
781 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers