Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
Brand:  Nabertherm™ NA--0681K2
Additional Details : Weight : 348.00000kg
 View more versions of this product
Product Code. 17338049
  • £9869.79 / Each
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Specifications
Specifications
B500
850°C
1330 x 790 x 1440 mm
11000 W
60 L
500 x 350 x 350 mm
400 V