Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed. Oustanding temperature uniformity and practical details set very high quality benchmarks.
Brand:  Nabertherm™ HT0246VNA
Additional Details : Weight : 306.00000kg
 View more versions of this product
Product Code. 17347839
  • £19085.61 / Each
Estimated Shipment: 01-10-2024
to see stock.
Add to basket
Description
Description
  • Dual shell housing with fan cooling for low shell temperatures
  • Heating from both sides via molybdenum disilicide heating elements
  • High-quality fiber insulation backed by special insulation
  • Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
  • Long-life roof insulation with special suspension
  • Chain-guided parallel swivel door for defined opening and closing of the door
  • Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
  • Heating elements switched via thyristors
Specifications
Specifications
8 L
Molybdenum Disilicide Heating Elements and Fiber Insulation
640 x 740 x 1755 mm
8500 W
Up to 1600°C
300 x 150 x 150 mm
400 V