Learn More
Zeiss™ Stemi 508 Doc (phototube 0/100%, right path) Stereo Microscope
Compact Greenough, apochromatic stereo microscope with 8:1 zoom for biological education, labs and production environments.
Brand:  Zeiss™ 495009-0019-000
Additional Details : Weight : 10.95500kg
Product Code. 17372830
  • £4327.30 / 1 set
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
  • The Stemi 508 is compact, reliable and equipped with optics and mechanics designed for heavy workloads. With the large 36 mm object field you always keep the overview of your sample.
  • The 8:1 zoom then allows to bring details up to 50× magnification. Add interchangeable optics and observe an area of up to 122 mm, making Stemi 508 a top performer in its class.
  • Select from a range of apochromatic front optics and eyepieces to reach any magnification between 2× and 250×. Double your resolution or enjoy large working distances up to 287 mm without compromising optical quality.
  • The Stemi 508 offers better ergonomics than any other Greenough-type stereo microscope: The low viewing angle of 35° lets you keep a relaxed posture even after hours of work. With the Stemi 508 you observe and document your samples exactly as they are: rich in detail, sharp in focus and free from distortion or color fringes.
  • The Stemi 508 is your robust all-rounder for everyday lab work and industrial inspections: accurate, ergonomic – and always easy to use.
  • Choose from a wide range of different stands and accessories to get exactly what you need for your application. From compact stands to flexible and stable boom stands, from basic transmitted light to polarization contrast.
  • Whether your object needs special illumination or simply the brightest light – you have various fiberoptic light guides and LED illuminators to choose from. Position your specimen precisely by adding a gliding, tilting or rotating polarization stage.
  • Via interchangeable camera adapters, Stemi 508 can be combined with any ZEISS Axiocam camera and most DSLR cameras or camcorders. A c-mount adapter 0.5x for Axiocam cameras up to chip size 1/2 in. is always included.
Specifications
Specifications
Stand K, CL 1500 HAL and gooseneck light guide
Biological Education, Labs and Production Environments.